Product Details

SNP ID
rs147337556
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:99028180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGATCACACGTGAGTGGGCTTC[A/G]CGCGGACCCAAAGTAGAACAGTCGG
Phenotype
MIM: 605568
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927550 PubMed Links

Gene Details

Gene
LOC101927550
Gene Name
uncharacterized LOC101927550
There are no transcripts associated with this gene.

Gene
SMURF1
Gene Name
SMAD specific E3 ubiquitin protein ligase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199847.1 4931 UTR 3 NP_001186776.1
NM_020429.2 4931 UTR 3 NP_065162.1
NM_181349.2 4931 UTR 3 NP_851994.1
XM_017012457.1 4931 UTR 3 XP_016867946.1

View Full Product Details