Product Details
- SNP ID
-
rs147464718
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:95360033 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCCTATGAGAATTTTCCCATGGTA[A/C]ACAGAAGCCACAGAGGTGCCCTGAA
- Phenotype
-
MIM: 602720
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
PON3
PubMed Links
Gene Details
- Gene
- PON3
- Gene Name
- paraoxonase 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000940.2 |
1030 |
Silent Mutation |
|
|
NP_000931.1 |
View Full Product Details