Product Details

SNP ID
rs147555041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:27155013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACGGCGCCCTCGGGAGCTGCGG[A/C]GGCAGTCGGACCTTCGTCCTTATGC
Phenotype
MIM: 142954 MIM: 142951 MIM: 142950 MIM: 142956
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HOXA-AS3 PubMed Links

Gene Details

Gene
HOXA-AS3
Gene Name
HOXA cluster antisense RNA 3
There are no transcripts associated with this gene.

Gene
HOXA10-HOXA9
Gene Name
HOXA10-HOXA9 readthrough
There are no transcripts associated with this gene.

Gene
HOXA3
Gene Name
homeobox A3
There are no transcripts associated with this gene.

Gene
HOXA6
Gene Name
homeobox A6
There are no transcripts associated with this gene.

Gene
HOXA7
Gene Name
homeobox A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006896.3 721 Missense Mutation GCC,TCC A197S NP_008827.2
Gene
HOXA9
Gene Name
homeobox A9
There are no transcripts associated with this gene.

View Full Product Details