Product Details

SNP ID
rs147595973
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6165770 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGTAATAGAGGCAGTTATCGGT[C/G]AGGATGAACCACCGGCGCTTCCAGG
Phenotype
MIM: 605081
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CYTH3 PubMed Links

Gene Details

Gene
CYTH3
Gene Name
cytohesin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004227.3 968 Silent Mutation CTC,CTG L288L NP_004218.1
Gene
USP42
Gene Name
ubiquitin specific peptidase 42
There are no transcripts associated with this gene.

View Full Product Details