Product Details

SNP ID
rs147887777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:141790525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATCCTGGGCCTGGCTGGCTGCC[A/G]ATTTCTCCTGCAGTGGCTGATCATT
Phenotype
MIM: 605062
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAS2R5 PubMed Links

Gene Details

Gene
TAS2R5
Gene Name
taste 2 receptor member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018980.2 309 Missense Mutation CAA,CGA Q55R NP_061853.1

View Full Product Details