Product Details

SNP ID
rs148251003
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:31338945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTGTCCAGAGCGTCGTTGAGGCC[A/G]TGCATCCTGTTCCTCTCGCGCGCGT
Phenotype
MIM: 611513
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
NEUROD6 PubMed Links

Gene Details

Gene
NEUROD6
Gene Name
neuronal differentiation 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022728.3 677 Silent Mutation CAC,CAT H108H NP_073565.2

View Full Product Details