Product Details

SNP ID
rs148344019
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:107768503 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGCAAAACTCTGGCTGAGTAATAA[A/G]AAAGATCAAAAGATAAATTAGACCT
Phenotype
MIM: 606872 MIM: 126650
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CBLL1 PubMed Links

Gene Details

Gene
CBLL1
Gene Name
Cbl proto-oncogene like 1
There are no transcripts associated with this gene.

Gene
SLC26A3
Gene Name
solute carrier family 26 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000111.2 Intron NP_000102.1
XM_011515867.2 Intron XP_011514169.1

View Full Product Details