Product Details

SNP ID
rs148513430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:122995170 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTTCTTGGTAGATGCTCCATGGT[A/G]AGTAACTGAATTTGAATGTAATTCC
Phenotype
MIM: 604867
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAS2R16 PubMed Links

Gene Details

Gene
TAS2R16
Gene Name
taste 2 receptor member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016945.2 531 Silent Mutation CTC,CTT L155L NP_058641.1

View Full Product Details