Product Details

SNP ID
rs148552166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:140407795 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACACTGAGACACAGCAGAACACGA[C/T]TGGAGTGGACTTTACCGTGCGTTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB19 PubMed Links

Gene Details

Gene
RAB19
Gene Name
RAB19, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008749.2 543 Missense Mutation ACT,ATT T50I NP_001008749.2
XM_005249996.3 543 Missense Mutation ACT,ATT T50I XP_005250053.1
XM_005249997.3 543 Missense Mutation ACT,ATT T50I XP_005250054.1
XM_005249998.3 543 Missense Mutation ACT,ATT T50I XP_005250055.1
XM_017012207.1 543 Missense Mutation ACT,ATT T50I XP_016867696.1
Gene
SLC37A3
Gene Name
solute carrier family 37 member 3
There are no transcripts associated with this gene.

View Full Product Details