Product Details
- SNP ID
-
hCV173361528
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:98851534 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGAAGGTGTGGGTCAGGTTGTAC[A/G]CTGTGCTGGCCACGGACACAGGGTG
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM130
PubMed Links
Gene Details
- Gene
- TMEM130
- Gene Name
- transmembrane protein 130
There are no transcripts associated with this gene.
View Full Product Details