Product Details
- SNP ID
-
rs148704543
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:30964074 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGCCACTGCTGGGCTCACCATGGA[C/T]CGCCGGATGTGGGGGGCCCACGTCT
- Phenotype
-
MIM: 139191
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GHRHR
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2302021] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GHRHR
- Gene Name
- growth hormone releasing hormone receptor
View Full Product Details