Product Details

SNP ID
rs148790247
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:80745135 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCCAGGTCCATGGGGACCATTTG[C/T]CCGTCACAACAGCCACCATTTCTGA
Phenotype
MIM: 602645
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEMA3C PubMed Links

Gene Details

Gene
SEMA3C
Gene Name
semaphorin 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006379.3 2577 Missense Mutation GAC,GGC D672G NP_006370.1
XM_005250113.2 2577 Missense Mutation GAC,GGC D614G XP_005250170.1

View Full Product Details