Product Details

SNP ID
rs149099078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:152648889 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGATGAGCTCGAGGCTTTCTGC[A/G]TTATAGTTTGTGTCGTTGCAAAAAG
Phenotype
MIM: 600375
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XRCC2 PubMed Links

Gene Details

Gene
XRCC2
Gene Name
X-ray repair cross complementing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005431.1 682 Missense Mutation NP_005422.1

View Full Product Details