Product Details
- SNP ID
-
rs149223563
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:100401299 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCCCTCTGAAAAGCTGGCTGCCAA[A/G]GCCTTGCTCTGGGGAGCTTTAAATT
- Phenotype
-
MIM: 605341
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PILRA
PubMed Links
Gene Details
- Gene
- PILRA
- Gene Name
- paired immunoglobin like type 2 receptor alpha
There are no transcripts associated with this gene.
- Gene
- ZCWPW1
- Gene Name
- zinc finger CW-type and PWWP domain containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001258008.1 |
2476 |
Missense Mutation |
CCT,CTT |
P484L |
NP_001244937.1 |
NM_017984.4 |
2476 |
Silent Mutation |
GCC,GCT |
A554A |
NP_060454.3 |
XM_005250479.2 |
2476 |
Missense Mutation |
CCT,CTT |
P485L |
XP_005250536.1 |
XM_005250480.2 |
2476 |
Missense Mutation |
CCT,CTT |
P458L |
XP_005250537.1 |
XM_006716035.3 |
2476 |
Silent Mutation |
GCC,GCT |
A556A |
XP_006716098.1 |
XM_006716036.2 |
2476 |
Silent Mutation |
GCC,GCT |
A556A |
XP_006716099.1 |
XM_006716037.3 |
2476 |
Silent Mutation |
GCC,GCT |
A555A |
XP_006716100.1 |
XM_006716038.3 |
2476 |
Silent Mutation |
GCC,GCT |
A555A |
XP_006716101.1 |
XM_006716040.3 |
2476 |
Silent Mutation |
GCC,GCT |
A529A |
XP_006716103.1 |
XM_017012379.1 |
2476 |
Missense Mutation |
CCT,CTT |
P457L |
XP_016867868.1 |
XM_017012380.1 |
2476 |
Missense Mutation |
CCT,CTT |
P457L |
XP_016867869.1 |
View Full Product Details