Product Details

SNP ID
rs149245237
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:143292686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTGTCCCCTTTACAAGAAGCTC[C/T]GCCTGTCGACAGGTGAGAATTGATG
Phenotype
MIM: 600639 MIM: 616524
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASP2 PubMed Links

Gene Details

Gene
CASP2
Gene Name
caspase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001224.4 458 Missense Mutation CGC,TGC R124C NP_001215.1
NM_032982.3 458 Missense Mutation CGC,TGC R155C NP_116764.2
NM_032983.3 458 Intron NP_116765.2
Gene
LOC105375545
Gene Name
uncharacterized LOC105375545
There are no transcripts associated with this gene.

Gene
TMEM139
Gene Name
transmembrane protein 139
There are no transcripts associated with this gene.

View Full Product Details