Product Details

SNP ID
rs149278091
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:83366239 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCTGTGTAATCCACTTGCTTTC[A/G]ATTTTTATTTAGGTGAAAACATCTT
Phenotype
MIM: 608166
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEMA3E PubMed Links

Gene Details

Gene
SEMA3E
Gene Name
semaphorin 3E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178129.1 3660 UTR 3 NP_001171600.1
NM_012431.2 3660 UTR 3 NP_036563.1

View Full Product Details