Product Details

SNP ID
rs149352998
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:19145297 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGAGAGGGCGTCTCGGGGAGGC[C/T]AGGGACAGGTCTGCGACGAAGTCCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FERD3L PubMed Links

Gene Details

Gene
FERD3L
Gene Name
Fer3-like bHLH transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152898.2 125 Silent Mutation CTA,CTG L22L NP_690862.1

View Full Product Details