Product Details

SNP ID
rs149477068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:6463144 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGTGATGGTCTTTGGCACTTA[C/T]GCTGGCAAACTGAGCTTCTTTCCCT
Phenotype
MIM: 609024
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDELR2 PubMed Links

Gene Details

Gene
KDELR2
Gene Name
KDEL endoplasmic reticulum protein retention receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100603.1 544 Missense Mutation CAT,CGT H128R NP_001094073.1
NM_006854.3 544 Silent Mutation GCA,GCG A212A NP_006845.1

View Full Product Details