Product Details

SNP ID
hCV173413481
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73444070 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGCTTCTTACCCGGGCGCCGGC[C/G]TGACCTTGCTGCAGGGGGGATGACG
Phenotype
MIM: 605681 MIM: 601766
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BAZ1B PubMed Links

Gene Details

Gene
BAZ1B
Gene Name
bromodomain adjacent to zinc finger domain 1B
There are no transcripts associated with this gene.

Gene
FZD9
Gene Name
frizzled class receptor 9
There are no transcripts associated with this gene.

View Full Product Details