Product Details

SNP ID
rs149748427
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100389962 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCAGTAGCACAACCACCACAACC[A/G]GCCTCAGGGTCACACAGGGCAAACG
Phenotype
MIM: 605341
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PILRA PubMed Links

Gene Details

Gene
PILRA
Gene Name
paired immunoglobin like type 2 receptor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013439.2 741 Missense Mutation AGC,GGC S177G NP_038467.2
NM_178272.1 741 Intron NP_840056.1
NM_178273.1 741 Intron NP_840057.1

View Full Product Details