Product Details

SNP ID
rs149935648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73593934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCTCAGGGATGCGGCCCGGGTC[A/G]GTCAGGATACTGGTAGATGTGCCCA
Phenotype
MIM: 605678
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MLXIPL PubMed Links

Gene Details

Gene
MLXIPL
Gene Name
MLX interacting protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032951.2 2538 Silent Mutation NP_116569.1
NM_032952.2 2538 Silent Mutation NP_116570.1
NM_032953.2 2538 Silent Mutation NP_116571.1
NM_032954.2 2538 Silent Mutation NP_116572.1
XM_011516277.1 2538 Silent Mutation XP_011514579.1
XM_011516278.1 2538 Silent Mutation XP_011514580.1
XM_011516279.1 2538 Silent Mutation XP_011514581.1
XM_011516281.2 2538 Silent Mutation XP_011514583.1
XM_017012263.1 2538 Silent Mutation XP_016867752.1

View Full Product Details