Product Details

SNP ID
rs149936159
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:112767462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACACCATATACTGCTTTCACTGTG[C/T]GTCCCTTTTCAGTCCAGCAGATGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM168 PubMed Links

Gene Details

Gene
TMEM168
Gene Name
transmembrane protein 168
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287497.1 2221 Missense Mutation CAC,CGC H610R NP_001274426.1
NM_022484.5 2221 Missense Mutation CAC,CGC H610R NP_071929.3
XM_017012523.1 2221 Missense Mutation CAC,CGC H610R XP_016868012.1
XM_017012524.1 2221 Intron XP_016868013.1
XM_017012525.1 2221 Missense Mutation CAC,CGC H226R XP_016868014.1

View Full Product Details