Product Details

SNP ID
rs150013489
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:77196636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACGATATTTGAGAAACTCATTAGC[A/C]ACATAAAACTGATCATACAAGGCAT
Phenotype
MIM: 605351
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CCDC146 PubMed Links

Gene Details

Gene
CCDC146
Gene Name
coiled-coil domain containing 146
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020879.2 1003 Intron NP_065930.2
Gene
FGL2
Gene Name
fibrinogen like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006682.2 1003 Silent Mutation GTG,GTT V321V NP_006673.1

View Full Product Details