Product Details

SNP ID
rs150017267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:25134908 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTATTATTTCACCATGTAAACTTT[A/T]ACTTCCTGTCATAATGTAACTAGTG
Phenotype
MIM: 616071 MIM: 123970
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C7orf31 PubMed Links

Gene Details

Gene
C7orf31
Gene Name
chromosome 7 open reading frame 31
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138811.3 2289 UTR 3 NP_620166.3
XM_005249619.3 2289 UTR 3 XP_005249676.1
XM_005249620.3 2289 UTR 3 XP_005249677.1
XM_011515125.2 2289 UTR 3 XP_011513427.1
Gene
CYCS
Gene Name
cytochrome c, somatic
There are no transcripts associated with this gene.

View Full Product Details