Product Details

SNP ID
rs150052347
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:4860075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGAACTTTCATTGGTATTCACC[C/T]GTTGCAAGGAAAATTCAGTAGCCTG
Phenotype
MIM: 607436 MIM: 611491
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PAPOLB PubMed Links

Gene Details

Gene
PAPOLB
Gene Name
poly(A) polymerase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020144.4 1920 Missense Mutation CAG,CGG Q579R NP_064529.4
Gene
RADIL
Gene Name
Rap associating with DIL domain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018059.4 1920 Intron NP_060529.4

View Full Product Details