Product Details

SNP ID
rs150275306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2524738 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGAAGATGAGGCCCTGGCCAGGC[A/G]CACGGGTGAGCCCTGGACTTGGGGC
Phenotype
MIM: 602576
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LFNG PubMed Links

Gene Details

Gene
LFNG
Gene Name
LFNG O-fucosylpeptide 3-beta-N-acetylglucosaminyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040167.1 493 Missense Mutation CAC,CGC H159R NP_001035257.1
NM_001040168.1 493 Missense Mutation CAC,CGC H159R NP_001035258.1
NM_001166355.1 493 Missense Mutation CAC,CGC H88R NP_001159827.1
NM_002304.2 493 Missense Mutation CAC,CGC H30R NP_002295.1
Gene
MIR4648
Gene Name
microRNA 4648
There are no transcripts associated with this gene.

View Full Product Details