Product Details

SNP ID
rs150300229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:36854930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGGACCTGTGGACTGTTTGGGA[A/C]ATTTATGCTAGGGAGGATCACGAGG
Phenotype
MIM: 606420
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ELMO1 PubMed Links

Gene Details

Gene
ELMO1
Gene Name
engulfment and cell motility 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039459.2 4579 UTR 3 NP_001034548.1
NM_001206480.2 4579 UTR 3 NP_001193409.1
NM_001206482.1 4579 UTR 3 NP_001193411.1
NM_014800.10 4579 UTR 3 NP_055615.8
NM_130442.3 4579 UTR 3 NP_569709.1
XM_005249919.2 4579 UTR 3 XP_005249976.1
XM_006715805.1 4579 UTR 3 XP_006715868.1
XM_011515654.1 4579 UTR 3 XP_011513956.1
XM_017012838.1 4579 UTR 3 XP_016868327.1
XM_017012839.1 4579 UTR 3 XP_016868328.1

View Full Product Details