Product Details

SNP ID
rs150465692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:55807229 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTACAAAGAAGCATATCTCGGAGCT[C/T]AACGAAGTCACAGTGATTTTCATTT
Phenotype
MIM: 612140
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEPT14 PubMed Links

Gene Details

Gene
SEPT14
Gene Name
septin 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207366.2 948 Missense Mutation AAG,GAG K283E NP_997249.2
XM_011515373.2 948 Missense Mutation AAG,GAG K283E XP_011513675.1

View Full Product Details