Product Details

SNP ID
rs150637497
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:93889120 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTCCGGTGACACCCACCGGAAAA[G/T]AATTTTTCACATGTCATGGAACTTA
Phenotype
MIM: 600033
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC105375401 PubMed Links

Gene Details

Gene
LOC105375401
Gene Name
uncharacterized LOC105375401
There are no transcripts associated with this gene.

Gene
TFPI2
Gene Name
tissue factor pathway inhibitor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271003.1 688 Missense Mutation TTA,TTC L114F NP_001257932.1
NM_001271004.1 688 Missense Mutation TTA,TTC L125F NP_001257933.1
NM_006528.3 688 Missense Mutation TTA,TTC L125F NP_006519.1

View Full Product Details