Product Details

SNP ID
rs150951637
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:75771899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGAAATACTCACATCACAGCCC[A/G]CTGGGAGCAGCTGTTACTGGTGAAT
Phenotype
MIM: 604697
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCL26 PubMed Links

Gene Details

Gene
CCL26
Gene Name
C-C motif chemokine ligand 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006072.4 372 Missense Mutation CGG,TGG R60W NP_006063.1
XM_017011671.1 372 Missense Mutation CGG,TGG R114W XP_016867160.1
XM_017011672.1 372 Missense Mutation CGG,TGG R60W XP_016867161.1

View Full Product Details