Product Details

SNP ID
rs151058099
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:100474281 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACGAAGCCCACCTACCTATCAT[C/T]GTCGCTGTCTAGGTGACCACTGATG
Phenotype
MIM: 615477 MIM: 611914
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NYAP1 PubMed Links

Gene Details

Gene
NYAP1
Gene Name
neuronal tyrosine phosphorylated phosphoinositide-3-kinase adaptor 1
There are no transcripts associated with this gene.

Gene
TSC22D4
Gene Name
TSC22 domain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303043.1 1677 Missense Mutation AAT,GAT N308D NP_001289972.1
NM_030935.4 1677 Missense Mutation AAT,GAT N308D NP_112197.1

View Full Product Details