Product Details

SNP ID
rs151103619
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:73313180 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCCGGCCACGGGCAGGGGTACC[C/T]GGGGGCAGGCAAAGGCTTCGTACAC
Phenotype
MIM: 615732 MIM: 612548
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NSUN5 PubMed Links

Gene Details

Gene
NSUN5
Gene Name
NOP2/Sun RNA methyltransferase family member 5
There are no transcripts associated with this gene.

Gene
TRIM50
Gene Name
tripartite motif containing 50
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281450.1 1331 Missense Mutation CAG,CGG Q401R NP_001268379.1
NM_001281451.1 1331 Missense Mutation CAG,CGG Q402R NP_001268380.1
NM_178125.3 1331 Missense Mutation CAG,CGG Q402R NP_835226.2
XM_011515787.1 1331 Intron XP_011514089.1
XM_011515788.2 1331 Missense Mutation CAG,CGG Q193R XP_011514090.1
XM_011515789.2 1331 Missense Mutation CAG,CGG Q401R XP_011514091.2

View Full Product Details