Product Details

SNP ID
rs111312995
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:6811622 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGGATGTGGCCACTTCTGCAGTG[A/G]CGCTGAAGTACAACGTGGAACTCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
XKR5 PubMed Links

Gene Details

Gene
XKR5
Gene Name
XK related 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289973.1 2207 Missense Mutation GCC,GTC A383V NP_001276902.1
NM_207411.4 2207 Missense Mutation GCC,GTC A546V NP_997294.3
XM_011534750.2 2207 Missense Mutation GCC,GTC A276V XP_011533052.1
XM_017013401.1 2207 Missense Mutation GCC,GTC A548V XP_016868890.1
XM_017013402.1 2207 Missense Mutation GCC,GTC A478V XP_016868891.1

View Full Product Details