Product Details
- SNP ID
-
rs111462156
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:10765680 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTCGGCCCTCTCGGGCTTTCCCTC[C/T]GTGTGCCTCTTGGCCTTAGGCTGGA
- Phenotype
-
MIM: 606505
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC102723313
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1078543] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC102723313
- Gene Name
- uncharacterized LOC102723313
There are no transcripts associated with this gene.
- Gene
- PINX1
- Gene Name
- PIN2/TERF1 interacting, telomerase inhibitor 1
There are no transcripts associated with this gene.
View Full Product Details