Product Details

SNP ID
rs111541291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:63017624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGGACACCATATACTGGATAC[C/T]TATATCCTGTAAGAAGAACACAAAT
Phenotype
MIM: 601509
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GGH PubMed Links

Gene Details

Gene
GGH
Gene Name
gamma-glutamyl hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003878.2 987 Missense Mutation AAG,AGG K235R NP_003869.1
XM_011517623.2 987 Intron XP_011515925.1

View Full Product Details