Product Details

SNP ID
rs111586207
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:26293766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAGAAAGCGTTTAATGCAAAATT[G/T]GGATATATAATTTTTGCAGAAATGT
Phenotype
MIM: 604941
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PPP2R2A PubMed Links

Gene Details

Gene
PPP2R2A
Gene Name
protein phosphatase 2 regulatory subunit Balpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001177591.1 Intron NP_001171062.1
NM_002717.3 Intron NP_002708.1
XM_005273559.1 Intron XP_005273616.1
XM_005273560.1 Intron XP_005273617.1
XM_011544565.1 Intron XP_011542867.1
XM_011544566.1 Intron XP_011542868.1
XM_011544567.2 Intron XP_011542869.1
XM_011544568.1 Intron XP_011542870.1
XM_011544569.1 Intron XP_011542871.1

View Full Product Details