Product Details

SNP ID
rs111835070
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:133238997 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCGGCTGCGGGTGCCCTCGCTGG[C/T]GTGGGAGCGGCTTCGGGTGCCCTCG
Phenotype
MIM: 605262 MIM: 603398
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NDRG1 PubMed Links

Gene Details

Gene
NDRG1
Gene Name
N-myc downstream regulated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135242.1 1149 Missense Mutation ACC,GCC T356A NP_001128714.1
NM_001258432.1 1149 Missense Mutation ACC,GCC T290A NP_001245361.1
NM_001258433.1 1149 Missense Mutation ACC,GCC T275A NP_001245362.1
NM_006096.3 1149 Missense Mutation ACC,GCC T356A NP_006087.2
XM_011516791.1 1149 Missense Mutation ACC,GCC T373A XP_011515093.1
XM_011516792.1 1149 Missense Mutation ACC,GCC T167A XP_011515094.1
Gene
WISP1
Gene Name
WNT1 inducible signaling pathway protein 1
There are no transcripts associated with this gene.

View Full Product Details