Product Details

SNP ID
rs112057667
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:96243723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGACTACATTAGATTTGTTGGCC[G/T]GATTCCAACAATCAGAACTAGGACT
Phenotype
MIM: 616930 MIM: 191330
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MTERF3 PubMed Links

Gene Details

Gene
MTERF3
Gene Name
mitochondrial transcription termination factor 3
There are no transcripts associated with this gene.

Gene
UQCRB
Gene Name
ubiquinol-cytochrome c reductase binding protein
There are no transcripts associated with this gene.

View Full Product Details