Product Details

SNP ID
rs112076531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:72548602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTTATAGGGCCTTATAGGCCTG[A/C]TTCTTCCCAATCCAAGCCTAGATAC
Phenotype
MIM: 607738
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
KCNB2 PubMed Links

Gene Details

Gene
KCNB2
Gene Name
potassium voltage-gated channel subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004770.2 Intron NP_004761.2
XM_017013981.1 Intron XP_016869470.1
XM_017013982.1 Intron XP_016869471.1

View Full Product Details