Product Details

SNP ID
rs112381660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:29069721 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGGCAGACGACGATTGCGGCTC[A/G]CAAATGATTTCGGATTTTGAAAAAA
Phenotype
MIM: 607350
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMBOX1 PubMed Links

Gene Details

Gene
HMBOX1
Gene Name
homeobox containing 1
There are no transcripts associated with this gene.

Gene
KIF13B
Gene Name
kinesin family member 13B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015254.3 6371 UTR 3 NP_056069.2
XM_005273458.1 6371 UTR 3 XP_005273515.1
XM_011544457.1 6371 UTR 3 XP_011542759.1
XM_011544458.1 6371 UTR 3 XP_011542760.1
XM_011544459.2 6371 UTR 3 XP_011542761.1
XM_011544460.2 6371 UTR 3 XP_011542762.1
XM_011544461.1 6371 UTR 3 XP_011542763.1
XM_017013257.1 6371 Intron XP_016868746.1

View Full Product Details