Product Details

SNP ID
rs112493967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143791268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACAGCGCCGGGTGAGGGCCTGCC[A/C]AGAAGCACCAGAGCCACTTCTCCAT
Phenotype
MIM: 607733
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LOC642574 PubMed Links

Gene Details

Gene
LOC642574
Gene Name
IQ motif and ankyrin repeat domain-containing protein LOC642574
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017014127.1 4870 Intron XP_016869616.1
Gene
SCRIB
Gene Name
scribbled planar cell polarity protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015356.4 4870 Silent Mutation CTG,CTT L1596L NP_056171.3
NM_182706.4 4870 Silent Mutation CTG,CTT L1621L NP_874365.3

View Full Product Details