Product Details

SNP ID
rs112573107
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86576075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGTTTATCTTCATTTTCTTTT[C/T]GTTTATCTTCATTTTCTTTTCCTTC
Phenotype
MIM: 605080
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CNGB3 PubMed Links

Gene Details

Gene
CNGB3
Gene Name
cyclic nucleotide gated channel beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019098.4 2207 Missense Mutation CAA,CGA Q720R NP_061971.3
XM_011517138.2 2207 Missense Mutation CAA,CGA Q582R XP_011515440.1

View Full Product Details