Product Details
- SNP ID
-
rs112599495
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:143567392 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCAGCCCCGGGCACGGGCCAGCAT[C/G]GCCACCTGCTGAGCGGACACGTGCG
- Phenotype
-
MIM: 611552
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GSDMD
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs189847213] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GSDMD
- Gene Name
- gasdermin D
There are no transcripts associated with this gene.
- Gene
- MROH6
- Gene Name
- maestro heat like repeat family member 6
- Gene
- NAPRT
- Gene Name
- nicotinate phosphoribosyltransferase
There are no transcripts associated with this gene.
View Full Product Details