Product Details

SNP ID
rs112795035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:47962154 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGGTGTTAGGGGCACACCTGTG[A/G]GACAGAGGCCTGACCTGGGCTCTGC
Phenotype
MIM: 602638 MIM: 600899
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MCM4 PubMed Links

Gene Details

Gene
MCM4
Gene Name
minichromosome maintenance complex component 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005914.3 1279 Missense Mutation AGA,GGA R113G NP_005905.2
NM_182746.2 1279 Missense Mutation AGA,GGA R113G NP_877423.1
Gene
PRKDC
Gene Name
protein kinase, DNA-activated, catalytic polypeptide
There are no transcripts associated with this gene.

View Full Product Details