Product Details

SNP ID
rs112813219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144099701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTGAAGAAGCAGCCCCAGGCCTC[A/G]CCTGATGGGGCCAGGTGGGAAGCAG
Phenotype
MIM: 123980 MIM: 610210 MIM: 611885
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CYC1 PubMed Links

Gene Details

Gene
CYC1
Gene Name
cytochrome c1
There are no transcripts associated with this gene.

Gene
MAF1
Gene Name
MAF1 homolog, negative regulator of RNA polymerase III
There are no transcripts associated with this gene.

Gene
SHARPIN
Gene Name
SHANK associated RH domain interactor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030974.3 Intron NP_112236.3
XM_017013887.1 Intron XP_016869376.1
XM_017013888.1 Intron XP_016869377.1
Gene
WDR97
Gene Name
WD repeat domain 97
There are no transcripts associated with this gene.

View Full Product Details