Product Details
- SNP ID
-
rs113162538
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:23191813 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCTCTTCCATCCTCTCCAAGGCAT[C/T]CAGCAGGGTGTGGATCGAGGCGTTC
- Phenotype
-
MIM: 603611
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TNFRSF10A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2230229] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TNFRSF10A
- Gene Name
- TNF receptor superfamily member 10a
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003844.3 |
1394 |
Missense Mutation |
AAT,GAT |
N430D |
NP_003835.3 |
View Full Product Details