Product Details

SNP ID
rs113242266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:25421592 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTACCAGAGCTCCTTTCAGGTCT[C/T]GGAGGGGAGAACGTGGCTGGTGCGT
Phenotype
MIM: 616904 MIM: 152760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DOCK5 PubMed Links

Gene Details

Gene
DOCK5
Gene Name
dedicator of cytokinesis 5
There are no transcripts associated with this gene.

Gene
GNRH1
Gene Name
gonadotropin releasing hormone 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000825.3 1928 Missense Mutation CAA,CGA Q77R NP_000816.4
NM_001083111.1 1928 Missense Mutation CAA,CGA Q73R NP_001076580.1
Gene
KCTD9
Gene Name
potassium channel tetramerization domain containing 9
There are no transcripts associated with this gene.

View Full Product Details