Product Details

SNP ID
rs113375628
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:101689173 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACAAAAGAAGCTGAAGGCGTCAC[A/G]GAGGAAGGCGCATCAGACCGCACAG
Phenotype
MIM: 606722
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCALD PubMed Links

Gene Details

Gene
NCALD
Gene Name
neurocalcin delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040624.1 1087 UTR 3 NP_001035714.1
NM_001040625.1 1087 UTR 3 NP_001035715.1
NM_001040626.1 1087 UTR 3 NP_001035716.1
NM_001040627.1 1087 UTR 3 NP_001035717.1
NM_001040628.1 1087 UTR 3 NP_001035718.1
NM_001040629.1 1087 UTR 3 NP_001035719.1
NM_001040630.1 1087 UTR 3 NP_001035720.1
NM_032041.2 1087 UTR 3 NP_114430.2
XM_011517332.2 1087 UTR 3 XP_011515634.1
XM_011517333.2 1087 UTR 3 XP_011515635.1
XM_011517334.2 1087 UTR 3 XP_011515636.1
XM_011517335.2 1087 UTR 3 XP_011515637.1
XM_017013900.1 1087 Intron XP_016869389.1
XM_017013901.1 1087 UTR 3 XP_016869390.1
XM_017013902.1 1087 UTR 3 XP_016869391.1

View Full Product Details