Product Details

SNP ID
rs113571531
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:78531021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGACACCTACTAAGTACCCTTT[C/T]GCCGCCTACAAGAAAGCATTTATAT
Phenotype
MIM: 606059
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PKIA PubMed Links

Gene Details

Gene
PKIA
Gene Name
protein kinase (cAMP-dependent, catalytic) inhibitor alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006823.3 Intron NP_006814.1
NM_181839.2 Intron NP_862822.1
XM_017013620.1 Intron XP_016869109.1
XM_017013621.1 Intron XP_016869110.1
XM_017013622.1 Intron XP_016869111.1
Gene
PKIA-AS1
Gene Name
PKIA antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details