Product Details

SNP ID
rs114026601
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:8318346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGCGTGCCGCACAGCGCCTCCTC[C/T]GAGGTGCCCGGCTGCTGCACCAGCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGK223 PubMed Links

Gene Details

Gene
SGK223
Gene Name
homolog of rat pragma of Rnd2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080826.2 4704 Silent Mutation TCA,TCG S1343S NP_001074295.2
XM_005272369.4 4704 Silent Mutation TCA,TCG S1343S XP_005272426.2
XM_005272370.4 4704 Silent Mutation TCA,TCG S1343S XP_005272427.2
XM_011543813.2 4704 Intron XP_011542115.1

View Full Product Details